Assay Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145090.3 | NP_659558.1 | 5 - 6 | 621 | 94 | |
XM_006235731.2 | XP_006235793.1 | 5 - 6 | 721 | 94 | ||
XM_006235732.1 | XP_006235794.1 | 5 - 6 | 662 | 94 | ||
XM_006235733.2 | XP_006235795.1 | 4 - 5 | 930 | 94 | ||
XM_006235734.2 | XP_006235796.1 | 4 - 5 | 929 | 94 | ||
GenBank mRNA | BC070895.1 | - | 5 - 6 | 602 | 94 | |
DQ167810.1 | - | 3 - 4 | 338 | 94 | ||
DQ167811.1 | - | 3 - 4 | 338 | 94 | ||
FQ222295.1 | - | 5 - 6 | 644 | 94 | ||
FQ232032.1 | - | 5 - 6 | 642 | 94 | ||
FQ233265.1 | - | 5 - 6 | 642 | 94 | ||
U35776.1 | - | 5 - 6 | 621 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
