Assay Details
Gene Symbol: Clcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022198.2 | NP_071534.2 | 12 - 13 | 2445 | 146 | |
XM_006256828.2 | XP_006256890.1 | 12 - 13 | 2748 | 146 | ||
GenBank mRNA | BC092209.1 | - | 12 - 13 | 2445 | 146 | |
FQ210965.1 | - | 12 - 13 | 2484 | 146 | ||
Z36944.1 | - | 10 - 11 | 2153 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P808769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76872 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P808769 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P808769 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P808769 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P808769 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P313445 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P686038 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700815659 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702122954 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20296 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15398 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90846 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68408 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17144 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702122954 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700815659 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10933269 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933265 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933266 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933275 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933270 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933268 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933276 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933263 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933272 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933271 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933273 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933262 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933274 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10933259 | - | - |
Rat Genome 230 2.0 Array | 1379932_at | - | - |
Rat Genome 230 2.0 Array | 1379932_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369791_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369791_at | - | - |
Affymetrix HT Rat Focus Array | 1369791_at | - | - |
Affymetrix HT Rat Focus Array | 1379932_at | - | - |
Affymetrix HT Rat Focus Array | 1379932_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369791_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00001975.rn.1 | TC0X00000257.rn.1 | RTA TC |
Rat Expression Set 230 | 1369791_PM_at | - | - |
Rat Expression Set 230 | 1369791_at | - | - |
Rat Expression Set 230 | 1379932_at | - | - |
Rat Expression Set 230 | 1379932_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10933259 | - | - |
Rat Genome U34 Set | Z36944cds_at | - | - |
Rat Genome U34 Set | Z36944cds_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21545195 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373419 | - | - |
RatRef-12 Expression BeadChip | ILMN_52585 | - | - |
