Assay Details
Gene Symbol: Clcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022198.2 | NP_071534.2 | 3 - 4 | 389 | 125 | |
XM_006256828.2 | XP_006256890.1 | 3 - 4 | 692 | 125 | ||
GenBank mRNA | BC092209.1 | - | 3 - 4 | 389 | 125 | |
FQ210965.1 | - | 3 - 4 | 428 | 125 | ||
Z36944.1 | - | 1 - 2 | 97 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
