Assay Details
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199253.2 | NP_954862.2 | 6 - 7 | 1183 | 80 | |
GenBank mRNA | AY847775.1 | - | 6 - 7 | 1183 | 80 | |
BC133063.1 | - | 6 - 7 | 1114 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
