Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 2 - 3 | 168 | 62 | |
GenBank mRNA | BC158690.1 | - | 2 - 3 | 168 | 62 | |
DY314066.1 | - | 2 - 3 | 168 | 62 | ||
EV767503.1 | - | 1 - 2 | 126 | 62 | ||
FQ216026.1 | - | 2 - 3 | 161 | 62 | ||
FQ216320.1 | - | 2 - 3 | 144 | 62 | ||
FQ218910.1 | - | 2 - 3 | 159 | 62 | ||
FQ230458.1 | - | 2 - 3 | 141 | 62 | ||
FQ233169.1 | - | 2 - 3 | 145 | 62 | ||
FQ234620.1 | - | 2 - 3 | 166 | 62 | ||
FQ234801.1 | - | 2 - 3 | 145 | 62 | ||
FQ235247.1 | - | 2 - 3 | 145 | 62 | ||
Y00350.1 | - | 1 - 2 | 124 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
