Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 4 - 5 | 304 | 94 | |
GenBank mRNA | BC158690.1 | - | 4 - 5 | 304 | 94 | |
DY314066.1 | - | 4 - 5 | 304 | 94 | ||
EV767503.1 | - | 3 - 4 | 262 | 94 | ||
FQ216026.1 | - | 4 - 5 | 297 | 94 | ||
FQ216320.1 | - | 4 - 5 | 280 | 94 | ||
FQ218910.1 | - | 4 - 5 | 295 | 94 | ||
FQ233169.1 | - | 4 - 5 | 281 | 94 | ||
FQ234620.1 | - | 4 - 5 | 302 | 94 | ||
FQ234801.1 | - | 4 - 5 | 281 | 94 | ||
FQ235247.1 | - | 4 - 5 | 281 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
