Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 5 - 6 | 510 | 65 | |
GenBank mRNA | BC158690.1 | - | 5 - 6 | 510 | 65 | |
DY314066.1 | - | 5 - 6 | 510 | 65 | ||
EV767503.1 | - | 4 - 5 | 468 | 65 | ||
FQ216026.1 | - | 5 - 6 | 503 | 65 | ||
FQ216320.1 | - | 5 - 6 | 486 | 65 | ||
FQ218910.1 | - | 5 - 6 | 501 | 65 | ||
FQ230458.1 | - | 6 - 7 | 355 | 65 | ||
FQ233169.1 | - | 5 - 6 | 487 | 65 | ||
FQ234620.1 | - | 5 - 6 | 508 | 65 | ||
FQ234801.1 | - | 5 - 6 | 487 | 65 | ||
FQ235247.1 | - | 5 - 6 | 487 | 65 | ||
Y00350.1 | - | 4 - 5 | 466 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
