Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 7 - 8 | 809 | 75 | |
GenBank mRNA | BC158690.1 | - | 7 - 8 | 809 | 75 | |
BI282626.1 | - | 3 - 4 | 75 | |||
FQ216320.1 | - | 7 - 8 | 785 | 75 | ||
FQ218910.1 | - | 7 - 8 | 800 | 75 | ||
FQ230458.1 | - | 8 - 9 | 654 | 75 | ||
FQ233169.1 | - | 7 - 8 | 786 | 75 | ||
FQ234620.1 | - | 7 - 8 | 807 | 75 | ||
FQ235247.1 | - | 7 - 8 | 786 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
