Assay Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077651.1 | NP_001071119.1 | 2 - 3 | 352 | 152 | |
XM_006238591.2 | XP_006238653.1 | 2 - 3 | 513 | 152 | ||
XM_006238592.2 | XP_006238654.1 | 2 - 3 | 828 | 152 | ||
XM_006238593.2 | XP_006238655.1 | 2 - 3 | 261 | 152 | ||
XM_006238594.2 | XP_006238656.1 | 2 - 3 | 537 | 152 | ||
XM_006238595.2 | XP_006238657.1 | 2 - 3 | 512 | 152 | ||
XM_008763893.1 | XP_008762115.1 | 2 - 3 | 439 | 152 | ||
GenBank mRNA | BC158558.1 | - | 2 - 3 | 363 | 152 | |
S83320.1 | - | 2 - 3 | 352 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
