Assay Details
Gene Symbol: Bend5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108672.1 | NP_001102142.1 | 5 - 6 | 1274 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P133485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P133485 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P133485 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P133485 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P133485 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P525495 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68115 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10870991 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870990 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870989 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870993 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870992 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10870987 | - | - |
Rat Genome 230 2.0 Array | 1383422_at | - | - |
Rat Genome 230 2.0 Array | 1383422_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383422_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383422_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500008237.rn.1 | TC0500001207.rn.1 | RTA TC |
Rat Expression Set 230 | 1383422_PM_at | - | - |
Rat Expression Set 230 | 1383422_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10870987 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21617345 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372980 | - | - |
