Assay Details
Gene Symbol: Elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044275.1 | NP_001037740.1 | 2 - 3 | 197 | 119 | |
GenBank mRNA | BC085795.1 | - | 2 - 3 | 197 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
