Assay Details
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015006.1 | NP_001015006.1 | 1 - 2 | 69 | 69 | |
GenBank mRNA | BC092651.1 | - | 1 - 2 | 69 | 69 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC092651.1, NM_001015006.1 transcripts.
This assay is in the 5'UTR region of the BC092651.1, NM_001015006.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
