Assay Details
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015006.1 | NP_001015006.1 | 3 - 4 | 300 | 75 | |
GenBank mRNA | BC092651.1 | - | 3 - 4 | 300 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
