Assay Details
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031000.3 | NP_112262.1 | 8 - 9 | 965 | 115 | |
GenBank mRNA | BC059133.1 | - | 8 - 9 | 1244 | 115 | |
D10854.1 | - | 8 - 9 | 965 | 115 | ||
FQ212586.1 | - | 8 - 9 | 1205 | 115 | ||
FQ212779.1 | - | 8 - 9 | 1240 | 115 | ||
FQ213220.1 | - | 7 - 8 | 912 | 115 | ||
FQ213483.1 | - | 8 - 9 | 1236 | 115 | ||
FQ213491.1 | - | 8 - 9 | 1238 | 115 | ||
FQ213579.1 | - | 8 - 9 | 1208 | 115 | ||
FQ213776.1 | - | 8 - 9 | 1207 | 115 | ||
FQ214173.1 | - | 8 - 9 | 1208 | 115 | ||
FQ214441.1 | - | 8 - 9 | 984 | 115 | ||
FQ214642.1 | - | 1221 | 115 | |||
FQ214700.1 | - | 8 - 9 | 1181 | 115 | ||
FQ215620.1 | - | 8 - 9 | 1032 | 115 | ||
FQ215772.1 | - | 8 - 9 | 1208 | 115 | ||
FQ218383.1 | - | 8 - 9 | 1208 | 115 | ||
FQ218721.1 | - | 8 - 9 | 1207 | 115 | ||
FQ218729.1 | - | 8 - 9 | 1207 | 115 | ||
FQ220055.1 | - | 6 - 7 | 808 | 115 | ||
FQ226221.1 | - | 8 - 9 | 1240 | 115 | ||
FQ228131.1 | - | 8 - 9 | 1241 | 115 | ||
FQ228132.1 | - | 8 - 9 | 1240 | 115 | ||
FQ228842.1 | - | 8 - 9 | 1236 | 115 | ||
FQ233156.1 | - | 8 - 9 | 1065 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
