Assay Details
Gene Symbol: Mrpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004235.1 | NP_001004235.1 | 2 - 3 | 585 | 85 | |
GenBank mRNA | BC079328.1 | - | 2 - 3 | 585 | 85 | |
FQ215540.1 | - | 2 - 3 | 598 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
