Assay Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 15 - 16 | 1644 | 110 | |
NM_134472.3 | NP_608302.2 | 14 - 15 | 1529 | 110 | ||
XM_006238632.2 | XP_006238694.1 | 15 - 16 | 1696 | 110 | ||
XM_006238633.1 | XP_006238695.1 | 14 - 15 | 1460 | 110 | ||
XM_006238634.2 | XP_006238696.1 | 14 - 15 | 1992 | 110 | ||
XM_006238635.2 | XP_006238697.1 | 15 - 16 | 1869 | 110 | ||
XM_006238636.2 | XP_006238698.1 | 15 - 16 | 1865 | 110 | ||
GenBank mRNA | CO403678.1 | - | 2 - 3 | 298 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
