Assay Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 7 - 8 | 732 | 125 | |
NM_134472.3 | NP_608302.2 | 6 - 7 | 617 | 125 | ||
XM_006238632.2 | XP_006238694.1 | 7 - 8 | 784 | 125 | ||
XM_006238633.1 | XP_006238695.1 | 6 - 7 | 548 | 125 | ||
XM_006238634.2 | XP_006238696.1 | 6 - 7 | 1080 | 125 | ||
XM_006238635.2 | XP_006238697.1 | 7 - 8 | 957 | 125 | ||
XM_006238636.2 | XP_006238698.1 | 7 - 8 | 953 | 125 | ||
GenBank mRNA | CO575506.1 | - | 6 - 7 | 540 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
