Assay Details
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 11 - 12 | 1985 | 147 | |
XM_006238646.2 | XP_006238708.1 | 11 - 12 | 1931 | 147 | ||
XM_006238647.2 | XP_006238709.1 | 9 - 10 | 967 | 147 | ||
XM_008763993.1 | XP_008762215.1 | 11 - 12 | 2008 | 147 | ||
GenBank mRNA | BC081913.1 | - | 11 - 12 | 1985 | 147 | |
FQ212041.1 | - | 10 - 11 | 1925 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
