Assay Details
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 8 - 9 | 1637 | 123 | |
XM_006238646.2 | XP_006238708.1 | 8 - 9 | 1583 | 123 | ||
XM_006238647.2 | XP_006238709.1 | 6 - 7 | 619 | 123 | ||
XM_008763993.1 | XP_008762215.1 | 8 - 9 | 1660 | 123 | ||
GenBank mRNA | BC081913.1 | - | 8 - 9 | 1637 | 123 | |
FQ212041.1 | - | 7 - 8 | 1577 | 123 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
