Assay Details
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004270.1 | NP_001004270.1 | 3 - 4 | 431 | 72 | |
XM_006243124.1 | XP_006243186.1 | 4 - 5 | 1057 | 72 | ||
XM_006243125.1 | XP_006243187.1 | 4 - 5 | 718 | 72 | ||
XM_006243126.1 | XP_006243188.1 | 4 - 5 | 737 | 72 | ||
XM_006243127.2 | XP_006243189.1 | 4 - 5 | 681 | 72 | ||
XM_006243128.2 | XP_006243190.1 | 4 - 5 | 1057 | 72 | ||
GenBank mRNA | BC079451.1 | - | 3 - 4 | 431 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
