Assay Details
Gene Symbol: Rab11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031152.2 | NP_112414.1 | 1 - 2 | 110 | 86 | |
GenBank mRNA | BC085727.1 | - | 1 - 2 | 110 | 86 | |
FQ214239.1 | - | 1 - 2 | 174 | 86 | ||
FQ220492.1 | - | 1 - 2 | 182 | 86 | ||
FQ234527.1 | - | 1 - 2 | 112 | 86 | ||
M75153.1 | - | 1 - 2 | 45 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
