Assay Details
Gene Symbol: Hspa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024351.2 | NP_077327.1 | 3 - 4 | 486 | 133 | |
GenBank mRNA | BC061547.1 | - | 3 - 4 | 493 | 133 | |
BC098914.1 | - | 3 - 4 | 438 | 133 | ||
M11942.1 | - | 3 - 4 | 457 | 133 |
Gene Symbol: LOC689955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764620.1 | XP_008762842.1 | 1 | 488 | 133 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
