Assay Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013181.1 | NP_001013199.1 | 2 - 3 | 1432 | 137 | |
GenBank mRNA | AB568265.1 | - | 1 - 2 | 230 | 137 | |
AY781102.1 | - | 2 - 3 | 1432 | 137 | ||
AY781103.1 | - | 2 - 3 | 1432 | 137 | ||
AY781104.1 | - | 2 - 3 | 1423 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
