Assay Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019179.1 | NP_062052.1 | 4 - 5 | 534 | 134 | |
XM_008767420.1 | XP_008765642.1 | 4 - 5 | 1023 | 134 | ||
XM_008767421.1 | XP_008765643.1 | 5 - 6 | 579 | 134 | ||
GenBank mRNA | BC126093.1 | - | 4 - 5 | 548 | 134 | |
L12138.1 | - | 4 - 5 | 534 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
