Assay Details
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033298.1 | NP_150640.1 | 3 - 4 | 592 | 111 | |
XM_006245675.2 | XP_006245737.1 | 3 - 4 | 605 | 111 | ||
GenBank mRNA | AB037472.1 | - | 2 - 3 | 369 | 111 | |
BC079403.1 | - | 3 - 4 | 592 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
