Assay Details
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033298.1 | NP_150640.1 | 8 - 9 | 1271 | 122 | |
XM_006245675.2 | XP_006245737.1 | 8 - 9 | 1284 | 122 | ||
GenBank mRNA | AB037472.1 | - | 7 - 8 | 1048 | 122 | |
BC079403.1 | - | 8 - 9 | 1271 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
