Assay Details
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 7 - 8 | 596 | 78 | |
GenBank mRNA | FM116382.1 | - | 7 - 8 | 555 | 78 | |
FM118664.1 | - | 7 - 8 | 580 | 78 | ||
FQ216044.1 | - | 7 - 8 | 595 | 78 | ||
FQ216100.1 | - | 7 - 8 | 572 | 78 | ||
FQ224925.1 | - | 7 - 8 | 593 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
