Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 12 - 13 | 1602 | 128 | |
XM_006257114.1 | XP_006257176.1 | 12 - 13 | 1618 | 128 | ||
XM_343797.7 | XP_343798.2 | 11 - 12 | 1717 | 128 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
