Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 17 - 18 | 2193 | 72 | |
XM_006257114.1 | XP_006257176.1 | 17 - 18 | 2209 | 72 | ||
XM_343797.7 | XP_343798.2 | 16 - 17 | 2308 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
