Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 18 - 19 | 2306 | 144 | |
XM_006257114.1 | XP_006257176.1 | 18 - 19 | 2322 | 144 | ||
XM_343797.7 | XP_343798.2 | 17 - 18 | 2421 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
