Assay Details
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 24 - 25 | 3060 | 76 | |
XM_006257114.1 | XP_006257176.1 | 24 - 25 | 3076 | 76 | ||
XM_343797.7 | XP_343798.2 | 23 - 24 | 3175 | 76 | ||
GenBank mRNA | AF155826.1 | - | 2 - 3 | 297 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
