Assay Details
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037180.1 | NP_001032257.1 | 7 - 8 | 1077 | 72 | |
GenBank mRNA | BC107454.1 | - | 7 - 8 | 1077 | 72 | |
FQ216680.1 | - | 3 - 4 | 471 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
