Assay Details
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037180.1 | NP_001032257.1 | 8 - 9 | 1199 | 105 | |
GenBank mRNA | BC107454.1 | - | 8 - 9 | 1199 | 105 | |
FQ216680.1 | - | 4 - 5 | 593 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
