Assay Details
Gene Symbol: Slc27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053580.2 | NP_446032.2 | 11 - 12 | 1811 | 57 | |
XM_006252953.2 | XP_006253015.1 | 11 - 12 | 1849 | 57 | ||
XM_006252954.1 | XP_006253016.1 | 11 - 12 | 1797 | 57 | ||
XM_008771156.1 | XP_008769378.1 | 11 - 12 | 1736 | 57 | ||
XM_008771157.1 | XP_008769379.1 | 11 - 12 | 1895 | 57 | ||
GenBank mRNA | BC074014.1 | - | 11 - 12 | 1811 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
