Assay Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053310.1 | NP_445762.1 | 1 - 2 | 72 | 84 | |
XM_006252890.2 | XP_006252952.1 | 2 - 3 | 198 | 84 | ||
XM_006252891.2 | XP_006252953.1 | 2 - 3 | 195 | 84 | ||
XM_008771094.1 | XP_008769316.1 | 2 - 3 | 196 | 84 | ||
XM_008771095.1 | XP_008769317.1 | 2 - 3 | 196 | 84 | ||
GenBank mRNA | AB020879.2 | - | 1 - 2 | 72 | 84 | |
BC070922.1 | - | 2 - 3 | 181 | 84 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB020879.2, BC070922.1, NM_053310.1, XM_006252890.2, XM_006252891.2, XM_008771094.1, XM_008771095.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
