Assay Details
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053310.1 | NP_445762.1 | 2 - 3 | 222 | 103 | |
XM_006252890.2 | XP_006252952.1 | 3 - 4 | 348 | 103 | ||
XM_006252891.2 | XP_006252953.1 | 3 - 4 | 345 | 103 | ||
XM_008771094.1 | XP_008769316.1 | 3 - 4 | 346 | 103 | ||
XM_008771095.1 | XP_008769317.1 | 3 - 4 | 346 | 103 | ||
GenBank mRNA | AB020879.2 | - | 2 - 3 | 222 | 103 | |
BC070922.1 | - | 3 - 4 | 331 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
