Assay Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012678.2 | NP_036810.1 | 1 - 2 | 160 | 146 | |
GenBank mRNA | FQ225054.1 | - | 171 | 146 | ||
FQ225082.1 | - | 171 | 146 | |||
J02780.1 | - | 1 - 2 | 163 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
