Assay Details
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115023.1 | NP_001108495.1 | 1 - 2 | 141 | 129 | |
XM_008771088.1 | XP_008769310.1 | 1 - 2 | 210 | 129 | ||
GenBank mRNA | BC158855.1 | - | 1 - 2 | 141 | 129 | |
FQ216881.1 | - | 1 - 2 | 142 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
