Assay Details
Gene Symbol: Ddx49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115023.1 | NP_001108495.1 | 3 - 4 | 75 | ||
XM_008771088.1 | XP_008769310.1 | 3 - 4 | 419 | 75 | ||
GenBank mRNA | BC158855.1 | - | 3 - 4 | 75 | ||
FQ216881.1 | - | 3 - 4 | 351 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
