Assay Details
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013880.2 | NP_001013902.2 | 6 - 7 | 864 | 96 | |
GenBank mRNA | BC079011.1 | - | 4 - 5 | 868 | 96 | |
CO388828.1 | - | 1 - 2 | 114 | 96 | ||
FQ229915.1 | - | 1 - 2 | 160 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
