Assay Details
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013880.2 | NP_001013902.2 | 8 - 9 | 1244 | 97 | |
GenBank mRNA | BC079011.1 | - | 6 - 7 | 1248 | 97 | |
CO388828.1 | - | 3 - 4 | 494 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
