Assay Details
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252788.2 | XP_006252850.1 | 4 - 5 | 1978 | 64 | |
XM_006252789.2 | XP_006252851.1 | 5 - 6 | 2087 | 64 | ||
XM_008771174.1 | XP_008769396.1 | 4 - 5 | 2086 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
