Assay Details
Gene Symbol: Cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106603.1 | NP_001100073.1 | 2 - 3 | 804 | 109 | |
GenBank mRNA | FQ214319.1 | - | 2 - 3 | 174 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
