Assay Details
Gene Symbol: Mob1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033891.1 | NP_001029063.1 | 3 - 4 | 441 | 57 | |
GenBank mRNA | BC101879.1 | - | 3 - 4 | 441 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117281 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1010867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44149 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1010867 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117281 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1010867 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117281 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1010867 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117281 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1010867 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222679 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39086 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12955 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99783 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10718461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10718460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856708 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10718459 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10856707 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10856703 | - | - |
Rat Genome 230 2.0 Array | 1392961_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397372_at | - | - |
Rat Genome 230 2.0 Array | 1371998_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397372_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371998_at | - | - |
Rat Genome 230 2.0 Array | 1392961_at | - | - |
Affymetrix HT Rat Focus Array | 1371998_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371998_at | - | - |
Affymetrix HT Rat Focus Array | 1392961_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392961_at | - | - |
Affymetrix HT Rat Focus Array | 1397372_at | - | - |
Affymetrix HT Rat Focus Array | 1397372_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400008827.rn.1 | TC0400001245.rn.1 | RTA TC |
Rat Expression Set 230 | 1371998_at | - | - |
Rat Expression Set 230 | 1371998_PM_at | - | - |
Rat Expression Set 230 | 1392961_at | - | - |
Rat Expression Set 230 | 1397372_PM_at | - | - |
Rat Expression Set 230 | 1397372_at | - | - |
Rat Expression Set 230 | 1392961_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10856703 | - | - |
Rat Genome U34 Set | rc_AI176611_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22065839 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359894 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530521 | - | - |
RatRef-12 Expression BeadChip | ILMN_161164 | - | - |
