Assay Details
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012096.1 | NP_001012096.1 | 10 - 11 | 1039 | 99 | |
XM_006236828.2 | XP_006236890.1 | 11 - 12 | 1149 | 99 | ||
XM_006236829.2 | XP_006236891.1 | 11 - 12 | 1148 | 99 | ||
XM_006236830.2 | XP_006236892.1 | 10 - 11 | 1116 | 99 | ||
XM_006236831.2 | XP_006236893.1 | 11 - 12 | 946 | 99 | ||
GenBank mRNA | BC087064.1 | - | 10 - 11 | 1039 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
