Assay Details
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107865.1 | NP_001101335.1 | 2 - 3 | 309 | 82 | |
XM_006236673.2 | XP_006236735.1 | 2 - 3 | 573 | 82 | ||
XM_006236674.2 | XP_006236736.1 | 2 - 3 | 573 | 82 | ||
XM_006236675.2 | XP_006236737.1 | 2 - 3 | 573 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
