Assay Details
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057205.2 | NP_476553.1 | 16 - 17 | 1248 | 89 | |
XM_006236936.2 | XP_006236998.1 | 15 - 16 | 1231 | 89 | ||
GenBank mRNA | AF336829.1 | - | 16 - 17 | 1257 | 89 | |
BC081743.1 | - | 16 - 17 | 1248 | 89 | ||
FQ215637.1 | - | 6 - 7 | 365 | 89 | ||
FQ234407.1 | - | 15 - 16 | 1208 | 89 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
