Assay Details
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057205.2 | NP_476553.1 | 3 - 4 | 185 | 137 | |
XM_006236936.2 | XP_006236998.1 | 2 - 3 | 168 | 137 | ||
GenBank mRNA | AF336829.1 | - | 3 - 4 | 194 | 137 | |
BC081743.1 | - | 3 - 4 | 185 | 137 | ||
FQ234407.1 | - | 2 - 3 | 144 | 137 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
