Assay Details
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025422.1 | NP_001020593.1 | 11 - 12 | 1505 | 97 | |
XM_008763194.1 | XP_008761416.1 | 11 - 12 | 1549 | 97 | ||
GenBank mRNA | BC098060.1 | - | 11 - 12 | 1505 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
