Assay Details
Gene Symbol: Isy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014188.1 | NP_001014210.1 | 2 - 3 | 108 | 59 | |
GenBank mRNA | BC079119.1 | - | 2 - 3 | 108 | 59 | |
FQ218201.1 | - | 2 - 3 | 159 | 59 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079119.1, NM_001014188.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
